Invitrogen
Manufacturer Code:PA584949
Catalog # PIPA584949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DNAAF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CILD13 FLJ25330 LRRC50 ODA7 swt; dynein (axonemal) assembly factor 1; Dynein assembly factor 1, axonemal; dynein, axonemal, assembly factor 1; Leucine-rich repeat-containing protein 50; outer row dynein assembly 7 homolog; testicular tissue protein Li 110
Gene Aliases: CILD13; DNAAF1; LRRC50; ODA7
UniProt ID: (Human) Q8NEP3
Entrez Gene ID: (Human) 123872
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.