Invitrogen
Manufacturer Code:PA5113927
Catalog # PIPA5113927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human LRRC15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hLib; hLib leucine rich repeat containing 15 leucine-rich repeat protein induced by beta amyloid Leucine-rich repeat protein induced by beta-amyloid homolog Leucine-rich repeat-containing protein 15 LIB LRRC15; leucine-rich repeat protein induced by beta amyloid; Leucine-rich repeat protein induced by beta-amyloid homolog; Leucine-rich repeat-containing protein 15
Gene Aliases: LIB; LRRC15
UniProt ID: (Human) Q8TF66
Entrez Gene ID: (Human) 131578
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.