Invitrogen
Manufacturer Code:PA5109269
Catalog # PIPA5109269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within human ApoER2 500-750 aa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4932703M08Rik AA921429 AI848122 Apoer2 Apolipoprotein E receptor 2 HSZ75190 Low-density lipoprotein receptor-related protein 8 LR8B Lrp8 LRP-8 MCI1 Protein LR8B; ApoE receptor 2; Apolipoprotein E receptor 2; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; Low-density lipoprotein receptor-related protein 8; LRP-8
Gene Aliases: APOER2; HSZ75190; LRP-8; LRP8; MCI1
UniProt ID: (Human) Q14114
Entrez Gene ID: (Human) 7804
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.