Invitrogen
Manufacturer Code:PA116913
Catalog # 01-950-155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide between residues 863-963 of the human ApoER2 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ApoE receptor 2; Apolipoprotein E receptor 2; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; Low-density lipoprotein receptor-related protein 8; Low-density lipoprotein receptor-related protein 8 antibody Apolipoprotein E receptor 2 antibody.; LR8B; LRP-8; Protein LR8B
Gene Aliases: 4932703M08Rik; AA921429; AI848122; APOER2; HSZ75190; LR8B; LRP-8; LRP8; MCI1
UniProt ID: (Chicken) Q98931, (Human) Q14114
Entrez Gene ID: (Chicken) 396102, (Human) 7804, (Mouse) 16975
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.