Invitrogen
Manufacturer Code:PA589161
Catalog # PIPA589161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 20-150 of human LRP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: crooked; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; Low-density lipoprotein receptor-related protein 6; LRP-6
Gene Aliases: ADCAD2; C030016K15Rik; Cd; Gw; LRP6; ska26; ska<m26Jus>; skax26; STHAG7
UniProt ID: (Human) O75581, (Mouse) O88572
Entrez Gene ID: (Human) 4040, (Rat) 312781, (Mouse) 16974
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.