Invitrogen
Manufacturer Code:PA547145
Catalog # PIPA547145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human LRP-6 Ala20-Pro1368 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: low density lipoprotein receptor-related protein 6; Low-density lipoprotein receptor-related protein 6; LRP-6; LRP6 low density lipoprotein receptor-related protein 6 low density lipoprotein-related protein 6 ADCAD2 FLJ90062 FLJ90421 lpr6
Gene Aliases: ADCAD2; LRP6; STHAG7
UniProt ID: (Human) O75581
Entrez Gene ID: (Human) 4040
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.