Invitrogen
Manufacturer Code:PA513144
Catalog # PIPA513144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1538-1567 amino acids from the C-terminal region of human LRP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 5 LRP7; Low-density lipoprotein receptor-related protein 7; LRP-5; LRP-7
Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; mKIAA4142; OPPG; OPS; OPTA1; VBCH2
UniProt ID: (Human) O75197, (Mouse) Q91VN0
Entrez Gene ID: (Human) 4041, (Mouse) 16973
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.