Abnova
Manufacturer Code:PAB12699
Catalog # 89-116-030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids (KATLY) of human LRP5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C to -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of LRP5.
For Research Use Only
Protein Aliases: BMND1; EVR1; EVR4; HBM; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LR3; LRP-5; LRP-7; LRP7; OPPG; OPS; OPTA1; VBCH2
Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; OPPG; OPS; OPTA1; VBCH2
UniProt ID: (Human) O75197
Entrez Gene ID: (Human) 4041
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.