Invitrogen
Manufacturer Code:PA576470
Catalog # PIPA576470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human LRP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein 330; gp330; low density lipoprotein-related protein 2; Low-density lipoprotein receptor-related protein 2; Low-density lipoprotein receptor-related protein 2 LRP-2 Glycoprotein 330 gp330 Megalin; LRP-2; Megalin
Gene Aliases: AI315343; AW536255; b2b1625.2Clo; D230004K18Rik; Gp330; Lrp2; Megalin
UniProt ID: (Rat) P98158, (Mouse) A2ARV4
Entrez Gene ID: (Rat) 29216, (Mouse) 14725
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.