Novus Biologicals
Manufacturer Code:NBP185292
Catalog # NBP185292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GFTSMSDRPGKRCAAEGSSPLLLLPDNVRIRKYNLSSERFSEYLQDEEYIQAVDYDWDPEDIGLSVVYYTVRGEGSRFGAIKRAYIPNFESGRNNLVQEVDLKLKYVMQPDGIAVDWVGRHIYWSDVKNKRIEVAKLDGRYRKWLISTDL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium sensor protein; calcium sensor protein DBSlow-density lipoprotein receptor-related protein 2 EC 1.1.2.3 EC 3.4.21.9 Glycoprotein 330 GP330 Heymann nephritis antigen homolog low density lipoprotein receptor-related protein 2 LRP-2 megalin; Glycoprotein 330; gp330; Heymann nephritis antigen homolog; low density lipoprotein receptor-related protein 2; Low-density lipoprotein receptor-related protein 2; LRP-2; Megalin
Gene Aliases: DBS; GP330; LRP2
UniProt ID: (Human) P98164
Entrez Gene ID: (Human) 4036
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.