Invitrogen
Manufacturer Code:PA522034
Catalog # PIPA522034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 611 and 937 of LRP130 (Uniprot IDNo.P42704) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa leucine-rich protein; CLONE-23970 GP130 LRP130 LSFC 130 kDa leucine-rich protein LRP 130 leucine-rich PPR motif-containing protein mitochondrial mitochondrial leucine-rich PPR motif-containing protein; GP130; leucine rich protein LRP130; leucine-rich pentatricopeptide repeat containing; Leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; LRP 130; mitochondrial leucine-rich PPR motif-containing protein
Gene Aliases: 3110001K13Rik; C76645; CLONE-23970; GP130; LRP130; LRPPRC; LSFC
UniProt ID: (Human) P42704, (Mouse) Q6PB66
Entrez Gene ID: (Human) 10128, (Mouse) 72416
Molecular Function:
RNA binding protein
kinase
nucleic acid binding
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.