Invitrogen
Manufacturer Code:PA581212
Catalog # PIPA581212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Mouse LRP1/CD91 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2MR; A2MR APOER APR CD91 FLJ16451 IGFBP3R LRP MGC88725 TGFBR5; alpha 2-macroglobulin receptor; Alpha-2-macroglobulin receptor; CD91; lipoprotein receptor-related protein; Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; LRP-1; LRP-515; LRP-85; lrp1; LRPICD; Prolow-density lipoprotein receptor-related protein 1
Gene Aliases: A2mr; AI316852; b2b1554Clo; CD91; Lrp; Lrp1
UniProt ID: (Mouse) Q91ZX7
Entrez Gene ID: (Mouse) 16971
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.