Invitrogen
Manufacturer Code:PA5116584
Catalog # PIPA5116584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-270 of human LRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2MR; A2MR AI316852 alpha 2-macroglobulin receptor alpha-2-macroglobulin receptor APOER Apolipoprotein E receptor APR b2b1554Clo CD91 FLJ16451 IGFBP3R LDL receptor related protein 1 lipoprotein receptor-related protein low density lipoprotein receptor-related protein 1 low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor) Low-density lipoprotein receptor-related protein 1 515 kDa subunit Low-density lipoprotein receptor-related protein 1 85 kDa subunit Low-density lipoprotein receptor-related protein 1 intracellular domain Lrp LRP1 LRP-1 LRP1A LRP-515 LRP-85 LRPICD MGC88725 Prolow-density lipoprotein receptor-related protein 1 TbetaR-V/LRP-1/IGFBP-3 receptor TGFBR5 type V tgf-beta receptor; alpha 2-macroglobulin receptor; Alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; CD91; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; LRP-1; LRP-515; LRP-85; lrp1; LRPICD; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; type V tgf-beta receptor
Gene Aliases: A2MR; AI316852; APOER; APR; b2b1554Clo; CD91; IGFBP3R; LRP; LRP1; LRP1A; TGFBR5
UniProt ID: (Human) Q07954, (Mouse) Q91ZX7, (Rat) G3V928
Entrez Gene ID: (Human) 4035, (Mouse) 16971, (Rat) 299858
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.