R&D Systems
Manufacturer Code:MAB1505SP
Catalog # MAB1505SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 255302 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human LRP-6 Ala20-Pro1368 Accession # O75581 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADCAD2 FLJ90062 FLJ90421 low density lipoprotein receptor-related protein 6 low-density lipoprotein receptor-related protein 6 LRP6 LRP-6; low density lipoprotein receptor-related protein 6; Low-density lipoprotein receptor-related protein 6; LRP-6
Gene Aliases: ADCAD2; LRP6; STHAG7
UniProt ID: (Human) O75581
Entrez Gene ID: (Human) 4040
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.