R&D Systems
Manufacturer Code:MAB6646SP
Catalog # MAB6646SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 747619 |
Immunogen | <EM>E. coli</EM>-derived recombinant human LRP-5 Intracellular Domain Phe1422-Ser1615 Accession # O75197 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BMND1 BMND1OPTA1 EVR1 EVR4 HBM low density lipoprotein receptor-related protein 5 low density lipoprotein receptor-related protein 7 low-density lipoprotein receptor-related protein 5 LR3VBCH2 LRP5 LRP-5 LRP7 LRP7exudative vitreoretinopathy 1 OPPG OPS OPTA1 osteoporosis pseudoglioma syndrome VBCH2; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LRP-5; LRP-7
Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; OPPG; OPS; OPTA1; VBCH2
UniProt ID: (Human) O75197
Entrez Gene ID: (Human) 4041
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.