Novus Biologicals
Manufacturer Code:NB1001886
Catalog # NB1001886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ERVEKTTGDKRT corresponding to internal region according to NP_002326.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BMND1OPTA1 EVR1 EVR4 HBM low density lipoprotein receptor-related protein 5 low density lipoprotein receptor-related protein 7 low-density lipoprotein receptor-related protein 5 LR3VBCH2 LRP-5 LRP7exudative vitreoretinopathy 1 OPPG OPS osteoporosis pseudoglioma syndrome; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LRP-5; LRP-7
Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; OPPG; OPS; OPTA1; VBCH2
UniProt ID: (Human) O75197
Entrez Gene ID: (Human) 4041
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.