R&D Systems
Manufacturer Code:MAB6360
Catalog # MAB6360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 545503 |
Immunogen | <EM>E. coli</EM>-derived recombinant human LRP-1 Gln4449-ala4544 Accession # Q07954 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A2MR; A2MR Alpha-2-Macroglobulin Receptor APOER Apolipoprotein E receptor APR CD91 CD91 antigen EC 1.1.1.94 EC 3.4.21.9 IGFBP3R low density lipoprotein receptor-related protein 1 LRP LRP-1 LRP1A prolow-density lipoprotein receptor-related protein 1 TbetaR-V/LRP-1/IGFBP-3 receptor TGFBR5 type V tgf-beta receptor; Alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; CD91; low density lipoprotein receptor-related protein 1; Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; LRP-1; LRP-515; LRP-85; LRPICD; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; type V tgf-beta receptor
Gene Aliases: A2MR; APOER; APR; CD91; IGFBP3R; LRP; LRP1; LRP1A; TGFBR5
UniProt ID: (Human) Q07954
Entrez Gene ID: (Human) 4035
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.