Novus Biologicals
Manufacturer Code:NBP272521AF488
Catalog # NB272521A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-303 of human LRH-1/NR5A2 (NP_995582) produced in E.coli. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Alpha-1-fetoprotein transcription factor; Alpha-1-fetoprotein transcription factor B1-binding factor b1-binding factor hepatocyte transcription factor which activates enhancer IIof hepatitis B virus B1F B1F2 fetoprotein-alpha 1 (AFP) transcription factor FTFLRH1 FTZ-F1 FTZ-F1beta HB1F hB1F-2 hB1FCYP7A promoter-binding factor Hepatocytic transcription factor Liver receptor homolog 1 liver receptor homolog-1 LRH-1CPF nuclear receptor NR5A2 nuclear receptor subfamily 5 group A member 2 nuclear receptor subfamily 5 group A member 2; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; CYP7A promoter-binding factor; fetoprotein-alpha 1 (AFP) transcription factor; hB1F; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; liver nuclear receptor homolog-1 variant 2; Liver receptor homolog 1; LRH-1; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2
Gene Aliases: B1F; B1F2; CPF; FTF; FTZ-F1; FTZ-F1beta; hB1F-2; LRH-1; LRH1; NR5A2
UniProt ID: (Human) O00482
Entrez Gene ID: (Human) 2494
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.