Proteintech
Manufacturer Code:121191AP150UL
Catalog # 50-172-7728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LRDD Fusion Protein Ag2761 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine-rich and death domain containing; Leucine-rich repeat and death domain-containing protein; leucine-rich repeats and death domain containing; LRDD PIDD; p53 protein induced, with death domain; p53-induced death domain-containing protein 1; p53-induced protein with a death domain; PIDD-C; PIDD-CC; PIDD-N
Gene Aliases: 1200011D09Rik; AU042446; LRDD; PIDD; PIDD1
UniProt ID: (Human) Q9HB75, (Mouse) Q9ERV7
Entrez Gene ID: (Human) 55367, (Mouse) 57913, (Rat) 293625
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.