Invitrogen
Manufacturer Code:PA542710
Catalog # PIPA542710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LRCH4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine rich repeat neuronal 4; leucine rich repeat protein 4, neuronal; Leucine-rich neuronal protein; Leucine-rich repeat and calponin homology domain-containing protein 4; Leucine-rich repeat neuronal protein 4; leucine-rich repeats and calponin homology (CH) domain containing 4; Leucine-rich repeats and calponin homology (CH) domain containing 4 FLJ40101 FLJ46315 LRN LRRN1 LRRN4 PP14183 SAP25
Gene Aliases: 2810008P14Rik; 2900069C24Rik; AI558103; LRCH4; LRN; LRRN1; LRRN4; mFLJ00248; PP14183; SAP25
UniProt ID: (Human) O75427, (Mouse) Q921G6
Entrez Gene ID: (Dog) 489836, (Human) 4034, (Horse) 100067371, (Bovine) 527134, (Rabbit) 100358516, (Guinea pig) 100722259, (Rat) 360779, (Mouse) 231798
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.