Novus Biologicals
Manufacturer Code:NBP250444G
Catalog # NBP250444G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M34-P1F10 |
Immunogen | Peptide Sequence - RDQRSVLASA (amino acids 190 -199) |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.135 LCA14 lecithin retinol acyltransferase lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase) MGC33103 Phosphatidylcholine--retinol O-acyltransferase; Lecithin retinol acyltransferase; Phosphatidylcholine--retinol O-acyltransferase
Gene Aliases: LCA14; LRAT
UniProt ID: (Human) O95237
Entrez Gene ID: (Human) 9227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.