Invitrogen
Manufacturer Code:PA536972
Catalog # PIPA536972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the 148-193 of human LRAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lecithin retinol acyltransferase; lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase); Lecithin retinol acyltransferase Phosphatidylcholine--retinol O-acyltransferase Phosphatidylcholine-retinol O-acyltransferase; Phosphatidylcholine--retinol O-acyltransferase; Phosphatidylcholine-retinol-O-acyltransferase
Gene Aliases: 1300010A18Rik; AI449251; LCA14; LRAT
UniProt ID: (Human) O95237, (Mouse) Q9JI60, (Rat) Q9JI61
Entrez Gene ID: (Human) 9227, (Mouse) 79235, (Rat) 64047
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.