Invitrogen
Manufacturer Code:PA112727
Catalog # 01-674-854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to LPPRP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -80° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lipid phosphate phosphatase-related protein type 3; LPR3 PRG-2 PRG2; PAP-2-like protein 2; Phospholipid phosphatase-related protein type 3; Plasticity-related gene 2 protein; PRG-2
Gene Aliases: LPPR3; LPR3; PHP2; PLPPR3; PRG-2; PRG2
UniProt ID: (Human) Q6T4P5
Entrez Gene ID: (Human) 79948
Molecular Function:
hydrolase
phosphatase
pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.