Invitrogen
Manufacturer Code:PA570476
Catalog # PIPA570476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human LPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDLCQ11 LIPD Lipoprotein lipase; Lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); Lipoprotein lipase-like protein; LPL; Phospholipase A1
Gene Aliases: HDLCQ11; LIPD; LPL
UniProt ID: (Pig) P49923, (Human) P06858, (Bovine) P11151, (Sheep) Q29524, (Rat) Q06000
Entrez Gene ID: (Pig) 397537, (Human) 4023, (Bovine) 280843, (Dog) 403626, (Goat) 100860750, (Rabbit) 100340171, (Horse) 100055358, (Sheep) 443408, (Rat) 24539
Molecular Function: esterase hydrolase lipase storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.