Abnova
Manufacturer Code:PAB25949
Catalog # 89-127-581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids at N-terminus of human LPHN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor L2; Calcium-independent alpha-latrotoxin receptor 2; CIRL-2; CIRL2; CL2; latrophilin 2; Latrophilin homolog 1; Latrophilin-2; LEC1; Lectomedin-1; LPHH1
Gene Aliases: ADGRL2; CIRL2; CL2; KIAA0786; LEC1; LPHH1; LPHN2
UniProt ID: (Human) A5XEI2
Entrez Gene ID: (Human) 23266
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.