Invitrogen
Manufacturer Code:PA5113143
Catalog # PIPA5113143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Acyl-CoA:lysophosphatidylglycerol acyltransferase 1 protein (151-341AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; Acyl-CoA:lysophosphatidylglycerol acyltransferase 1 AI649174 AW112037 BC013667 FAM34A FAM34A1 family with sequence similarity 34 member A KIAA0205 Lpgat1 lysophosphatidylglycerol acyltransferase 1 NET8; Acyl-CoA:monoacylglycerol acyltransferase LPGAT1; family with sequence similarity 34, member A
Gene Aliases: FAM34A; FAM34A1; KIAA0205; LPGAT1; NET8
UniProt ID: (Human) Q92604
Entrez Gene ID: (Human) 9926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.