Novus Biologicals
Manufacturer Code:NBP2852220.1ML
Catalog # NB033151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human LPGAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; acyl-CoA:lysophosphatidylglycerol acyltransferase 1 EC 2.3.1.- FAM34A FAM34A1 family with sequence similarity 34 member A KIAA0205 lysophosphatidylglycerol acyltransferase 1 NET8; Acyl-CoA:monoacylglycerol acyltransferase LPGAT1; family with sequence similarity 34, member A
Gene Aliases: FAM34A; FAM34A1; KIAA0205; LPGAT1; NET8
UniProt ID: (Human) Q92604
Entrez Gene ID: (Human) 9926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.