Abnova
Manufacturer Code:H00010162K
Catalog # 89-209-888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human LPCAT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human LPCAT3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; C3F MBOAT5 OACT5 nessy; LPCAT; LPLAT 5; LPSAT; Lyso-PC acyltransferase 3; lyso-PS acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 3; Lysophosphatidylserine acyltransferase; Lysophospholipid acyltransferase 5; membrane bound O-acyltransferase domain containing 5; Membrane-bound O-acyltransferase domain-containing protein 5; O-acyltransferase (membrane bound) domain containing 5; O-acyltransferase domain-containing protein 5; putative protein similar to nessy
Gene Aliases: C3F; LPCAT; LPCAT3; LPLAT 5; LPSAT; MBOAT5; nessy; OACT5
UniProt ID: (Human) Q6P1A2
Entrez Gene ID: (Human) 10162
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.