Invitrogen
Manufacturer Code:PA5101406
Catalog # PIPA5101406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LPCAT2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase 11; 1-acylglycerol-3-phosphate O-acyltransferase 11 1-acylglycerophosphocholine O-acyltransferase 1-AGP acyltransferase 11 1-AGPAT 11 1-alkylglycerophosphocholine O-acetyltransferase A330042H22 Acetyl-CoA:lyso-PAF acetyltransferase acetyl-CoA:lyso-platelet-activating factor acetyltransferase acyl-CoA:lysophosphatidylcholine acyltransferase 2 acyl-CoA:lysophosphatidylcholine acyltransferase member 1 acyltransferase like 1 acyltransferase like 1A acyltransferase-like 1 acyltransferase-like 1-A AGPAT11 AYTL1 Aytl1a LPAAT-alpha lpafat1 LPC acyltransferase 2 Lpcat2 LPCAT-2 Lpcat2a lyso-PAF acetyltransferase lysoPAFAT lysoPAFAT/LPCAT2 lysoPC acyltransferase 2 lysophosphatidic acid acyltransferase alpha Lysophosphatidylcholine acyltransferase 2 lyso-platelet-activating factor (PAF) acetyltransferase RGD1563994; 1-acylglycerophosphocholine O-acyltransferase; 1-AGP acyltransferase 11; 1-AGPAT 11; 1-alkenylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; Acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholine acyltransferase 2; acyl-CoA:lysophosphatidylcholine acyltransferase member 1; acyltransferase like 1; acyltransferase like 1A; Acyltransferase-like 1; acyltransferase-like 1-A; LPAAT-alpha; LPC acyltransferase 2; LPCAT-2; lyso-PAF acetyltransferase; lyso-platelet-activating factor (PAF) acetyltransferase; lysoPAFAT; lysoPAFAT/LPCAT2; lysoPC acyltransferase 2; Lysophosphatidic acid acyltransferase alpha; Lysophosphatidylcholine acyltransferase 2
Gene Aliases: A330042H22; AGPAT11; AYTL1; Aytl1a; lpafat1; LPCAT-2; LPCAT2; Lpcat2a; LysoPAFAT; RGD1563994
UniProt ID: (Human) Q7L5N7, (Mouse) Q8BYI6
Entrez Gene ID: (Human) 54947, (Mouse) 270084, (Rat) 100359680
Molecular Function: acyltransferase annexin calcium-binding protein calmodulin intracellular calcium-sensing protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.