Invitrogen
Manufacturer Code:PA5106278
Catalog # PIPA5106278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LPCAT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase 1-alkylglycerophosphocholine O-acetyltransferase 2900035H07Rik Acetyl-CoA:lyso-PAF acetyltransferase acetyl-CoA:lyso-platelet-activating factor acetyltransferase acyl-CoA:lysophosphatidylcholin acyl-CoA:lysophosphatidylcholine acyltransferase 1 acyltransferase like 2 acyltransferase-like 2 AGPAT10 AGPAT9 AYTL2 BB137372 BC005662 C87117 LPC acyltransferase 1 LPCAT Lpcat1 LPCAT-1 lyso-PAF acetyltransferase lysoPAFAT lysoPC acyltransferase 1 lysophosphatidylcholine acyltransferase 1 mLPCAT1 PFAAP3 Phosphonoformate immuno-associated protein 3 rd11 regulated by phosphonoformate RGD1311599; 1-alkenylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; Acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; Acyltransferase-like 2; LPC acyltransferase 1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; Lysophosphatidylcholine acyltransferase 1; mLPCAT1; Phosphonoformate immuno-associated protein 3; regulated by phosphonoformate
Gene Aliases: 2900035H07Rik; AGPAT10; AGPAT9; AYTL2; BB137372; BC005662; C87117; LPCAT; LPCAT1; PFAAP3; rd11; RGD1311599
UniProt ID: (Human) Q8NF37, (Mouse) Q3TFD2, (Rat) Q1HAQ0
Entrez Gene ID: (Human) 79888, (Mouse) 210992, (Rat) 361467
Molecular Function:
acyltransferase
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.