Abnova
Manufacturer Code:PAB26072
Catalog # 89-127-712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids at internal region of human LPCAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of LPCAT1.
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-alkenylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; Acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; Acyltransferase-like 2; AYTL2; FLJ12443; FLJ41609; LPC acyltransferase 1; lpcat; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; Lysophosphatidylcholine acyltransferase 1; PFAAP3; Phosphonoformate immuno-associated protein 3; regulated by phosphonoformate
Gene Aliases: AGPAT10; AGPAT9; AYTL2; lpcat; LPCAT1; PFAAP3
UniProt ID: (Human) Q8NF37
Entrez Gene ID: (Human) 79888
Molecular Function: acyltransferase annexin calcium-binding protein calmodulin intracellular calcium-sensing protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.