Proteintech
Manufacturer Code:221651AP150UL
Catalog # 50-173-3753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LPAR4 Fusion Protein Ag17817 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 23 GPR23 LPA 4 LPA receptor 4 LPA4 LPAR4 P2RY9 P2Y purinoceptor 9 P2Y5 LIKE P2Y5 like receptor P2Y9 Purinergic receptor 9; G protein-coupled receptor 23; G-protein coupled receptor 23; LPA receptor 4; LPA-4; Lysophosphatidic acid receptor 4; P2Y purinoceptor 9; P2Y5-like receptor; P2Y9; Purinergic receptor 9
Gene Aliases: 5730485F04Rik; GPR23; LPA4; LPAR4; P2RY9; P2Y5-LIKE; P2Y9; RGD1563686
UniProt ID: (Human) Q99677, (Mouse) Q8BLG2
Entrez Gene ID: (Human) 2846, (Rat) 302378, (Mouse) 78134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.