Novus Biologicals
Manufacturer Code:NBP201354
Catalog # NBP201354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G6 |
Immunogen | Full length human recombinant protein of human LPAR1(NP_001392) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: edg-2 EDG2Lysophosphatidic acid receptor Edg-2 endothelial differentiation lysophosphatidic acid G-protein-coupled receptor2 Gpcr26 GPR26 LPA receptor 1 LPA-1 LPA1vzg-1 lysophosphatidic acid receptor 1 Mrec1.3 rec.1.3 ventricular zone gene 1 VZG1; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; LPA receptor 1; LPA-1; Lysophosphatidic acid receptor 1; Lysophosphatidic acid receptor Edg-2; ventricular zone gene 1
Gene Aliases: edg-2; EDG2; Gpcr26; GPR26; LPA1; LPAR1; Mrec1.3; rec.1.3; vzg-1; VZG1
UniProt ID: (Human) Q92633
Entrez Gene ID: (Human) 1902
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.