Novus Biologicals
Manufacturer Code:NBP130327B
Catalog # NBP130327B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300] |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lysyl oxidase; lysyl oxidaseEC 1.4.3.13 MGC105112 protein-lysine 6-oxidase; Protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form
Gene Aliases: LOX
UniProt ID: (Human) P28300
Entrez Gene ID: (Human) 4015
Molecular Function: hydrolase oxidase oxidoreductase protease receptor serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.