Novus Biologicals
Manufacturer Code:NBP181734
Catalog # NBP181734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VEEKIKQTHRKYLLQEQLKIIKKELGLEKDDKDAIEEKFRERLKELVVPKHVMDVVDEELSKLGLLDNHSSEFNVTRNYLDWLTSIPWGKYSNENLDLARAQAVLEEDHYGMEDVKKRILEFIAVSQLRGSTQGKILCFYGP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.- EC 3.4.21.53 hLON hLON ATP-dependent protease LON lon peptidase 1 mitochondrial lon protease homolog mitochondrial Lon protease-like protein LONHs LONP Mitochondrial ATP-dependent protease Lon mitochondrial lon peptidase 1 mitochondrial lon protease-like protein PIM1 protease serine 15 PRSS15MGC1498 Serine protease 15; hLON ATP-dependent protease; Lon protease homolog, mitochondrial; Lon protease-like protein; LONHs; LONP; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial lon protease-like protein; Serine protease 15
Gene Aliases: CODASS; hLON; LON; LonHS; LONP; LONP1; PIM1; PRSS15
UniProt ID: (Human) P36776
Entrez Gene ID: (Human) 9361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.