Proteintech
Manufacturer Code:154401AP150UL
Catalog # 50-556-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LONP1 Fusion Protein Ag7196 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hLON ATP-dependent protease; hLON LON lon peptidase 1 mitochondrial Lon protease like protein LonHS LONP LONP1 PIM1 PRSS15 Serine protease 15; Lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03120}; Lon protease-like protein; lon protease-like protein {ECO:0000255|HAMAP-Rule:MF_03120}; LONHs; LONP; LONP {ECO:0000255|HAMAP-Rule:MF_03120}; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255|HAMAP-Rule:MF_03120}; mitochondrial lon protease-like protein; protease, serine, 15; Serine protease 15; serine protease 15 {ECO:0000255|HAMAP-Rule:MF_03120}
Gene Aliases: 1200017E13Rik; CODASS; hLON; LON; LonHS; LONP; LONP1; PIM1; PRSS15
UniProt ID: (Human) P36776, (Mouse) Q8CGK3, (Rat) Q924S5
Entrez Gene ID: (Human) 9361, (Mouse) 74142, (Rat) 170916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.