Novus Biologicals
Manufacturer Code:NBP299860AF350
Catalog # NB089834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant LOK (NP_005981.3 Arg 18-Glu 317). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 LOKserine/threonine-protein kinase 10 Lymphocyte-oriented kinase PRO2729 serine/threonine kinase 10; Lymphocyte-oriented kinase; Serine/threonine-protein kinase 10
Gene Aliases: LOK; PRO2729; STK10
UniProt ID: (Human) O94804
Entrez Gene ID: (Human) 6793
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.