R&D Systems
Manufacturer Code:MAB5075
Catalog # MAB5075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 475417 |
Immunogen | <EM>E. coli</EM>-derived recombinant human LMW-PTP/ACP1 Ala2-His158 Accession # P24666 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 1 soluble acid phosphatase of erythrocyte ACP1 Adipocyte acid phosphatase cytoplasmic phosphotyrosyl protein phosphatase EC 3.1.3.2 EC 3.1.3.48 HAAP LMWPTP LMW-PTP LMW-PTPase Low molecular weight cytosolic acid phosphatase low molecular weight phosphotyrosine protein phosphatase MGC111030 MGC3499 protein tyrosine phosphatase Red cell acid phosphatase 1; acid phosphatase of erythrocyte; Adipocyte acid phosphatase; cytoplasmic phosphotyrosyl protein phosphatase; LMW-PTP; LMW-PTPase; Low molecular weight cytosolic acid phosphatase; Low molecular weight phosphotyrosine protein phosphatase; protein tyrosine phosphatase; Red cell acid phosphatase 1; testicular secretory protein Li 37
Gene Aliases: ACP1; HAAP; LMW-PTP
UniProt ID: (Human) P24666
Entrez Gene ID: (Human) 52
Molecular Function:
hydrolase
oxidoreductase
phosphatase
protein phosphatase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.