R&D Systems
Manufacturer Code:MAB7709SP
Catalog # MAB7709SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 792520 |
Immunogen | <EM>E. coli</EM>-derived recombinant human LMP2/PSMB9 Thr21-Glu219 Accession # P28065 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta1i LMP2MGC70470 Low molecular mass protein 2 Macropain chain 7 Multicatalytic endopeptidase complex chain 7 proteasome (prosome macropain) subunit beta type 9 (large multifunctionalpeptidase 2) proteasome catalytic subunit 1i Proteasome chain 7 proteasome subunit beta 6i proteasome subunit beta type-9 Proteasome subunit beta-1i proteasome-related gene 2 PSMB6iproteasome (prosome macropain) subunit beta type 9 (large multifunctionalprotease 2) PSMB9 Really interesting new gene 12 protein RING12 RING12EC 3.4.25.1; large multifunctional peptidase 2; Low molecular mass protein 2; Macropain chain 7; Multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome catalytic subunit 1i; Proteasome chain 7; proteasome subunit beta 6i; Proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; Really interesting new gene 12 protein
Gene Aliases: beta1i; LMP2; PSMB6i; PSMB9; RING12
UniProt ID: (Human) P28065
Entrez Gene ID: (Human) 5698
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.