Invitrogen
Manufacturer Code:PA599989
Catalog # PIPA599989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LMOD3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5430424A14Rik leiomodin 3 leiomodin 3 (fetal) leiomodin fetal form Leiomodin-3 Lmod3 NEM10 RGD1564924; leiomodin 3 (fetal); Leiomodin, fetal form; Leiomodin-3
Gene Aliases: 5430424A14Rik; LMOD3; NEM10; RGD1564924
UniProt ID: (Human) Q0VAK6, (Mouse) E9QA62
Entrez Gene ID: (Human) 56203, (Mouse) 320502, (Rat) 500267
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.