Proteintech
Manufacturer Code:149481AP150UL
Catalog # 50-172-9857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LMOD3 Fusion Protein Ag6758 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leiomodin 3 (fetal); Leiomodin 3 leiomodin 3 (fetal) Leiomodin fetal form LMOD3; Leiomodin, fetal form; Leiomodin-3
Gene Aliases: 5430424A14Rik; LMOD3; NEM10; RGD1564924
UniProt ID: (Human) Q0VAK6, (Mouse) E9QA62
Entrez Gene ID: (Human) 56203, (Mouse) 320502, (Rat) 500267
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.