Proteintech
Manufacturer Code:151171AP150UL
Catalog # 50-172-9994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LMOD1 Fusion Protein Ag7176 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1D 64 kDa autoantigen 1D 64 kDa autoantigen 1D3 64 kDa autoantigen D1 64kD D1 Leiomodin 1 leiomodin 1 (smooth muscle) Leiomodin muscle form LMOD1 SM LMOD Smooth muscle leiomodin; 64 kDa autoantigen 1D; 64 kDa autoantigen 1D3; 64 kDa autoantigen D1; leimodin 1 (smooth muscle); leiomodin 1 (smooth muscle); Leiomodin, muscle form; Leiomodin-1; SM-Lmod; Smooth muscle leiomodin; thyroid and eye muscle autoantigen D1 (64kD); Thyroid-associated ophthalmopathy autoantigen
Gene Aliases: 1D; 64kD; 9530015K06Rik; D1; LMOD1; SM-LMOD; SMLMOD
UniProt ID: (Human) P29536, (Rat) A0A0G2K0D3, (Mouse) Q8BVA4
Entrez Gene ID: (Human) 25802, (Rat) 304816, (Mouse) 93689
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.