Abnova
Manufacturer Code:MAB11034
Catalog # 89-109-248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP51 |
Immunogen | A synthetic peptide corresponding to near N-terminus of humann LMO2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit monoclonal antibody raised againstsynthetic peptide of LMO2.
For Research Use Only
Protein Aliases: Cysteine-rich protein TTG-2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LMO-2; RBTN2; RBTNL1; RHOM2; rhombotin 2; Rhombotin-2; rhombotin-like 1; T-cell translocation gene 2; T-cell translocation protein 2; TTG2
Gene Aliases: LMO2; RBTN2; RBTNL1; RHOM2; TTG2
UniProt ID: (Human) P25791
Entrez Gene ID: (Human) 4005
Molecular Function:
RNA binding protein
actin family cytoskeletal protein
cytoskeletal protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.