Proteintech
Manufacturer Code:219661AP150UL
Catalog # 50-173-3661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LMO2 Fusion Protein Ag16795 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteine rich protein TTG 2 LIM domain only protein 2 LMO 2 LMO2 RBTN2 RBTNL1 RHOM2 Rhombotin 2 T cell translocation protein 2 TTG2; Cysteine-rich protein TTG-2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LIM only 2; LMO-2; rhombotin 2; Rhombotin-2; rhombotin-like 1; T-cell translocation gene 2; T-cell translocation protein 2
Gene Aliases: LMO2; Rbtn-2; RBTN2; RBTNL1; Rhom-2; RHOM2; TTG2
UniProt ID: (Human) P25791, (Mouse) P25801
Entrez Gene ID: (Human) 4005, (Mouse) 16909
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.