Novus Biologicals
Manufacturer Code:NBP304860100UL
Catalog # NB093817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 220-300 of human LMBRD1 (NP_060838.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA810I22.1 C6orf209 chromosome 6 open reading frame 209 FLJ11240 FLJ11854 HDAg-L-interacting protein NESI hepatitis delta antigen-L interacting protein liver regeneration p-53 related protein LMBD1 LMBR1 domain containing 1 LMBR1 domain-containing protein 1 NESIcblF Nuclear export signal-interacting protein probable lysosomal cobalamin transporter; HDAg-L-interacting protein NESI; hepatitis delta antigen-L interacting protein; liver regeneration p-53 related protein; LMBD1; LMBR1 domain-containing protein 1; Lysosomal cobalamin transport escort protein LMBD1; Nuclear export signal-interacting protein
Gene Aliases: BM-021; C6orf209; CD001; LMBD1; LMBRD1; MAHCF; MSTP044; NESI
UniProt ID: (Human) Q9NUN5
Entrez Gene ID: (Human) 55788
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.