Novus Biologicals
Manufacturer Code:NBP271152V
Catalog # NBP271152V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A8 |
Immunogen | Full length recombinant protein of human LMAN1 (NP_005561) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: endoplasmic reticulum-golgi intermediate compartment protein 53; ER-Golgi intermediate compartment 53 kDa protein; ERGIC-53 ERGIC53ER-Golgi intermediate compartment 53 kDa protein F5F8Dendoplasmic reticulum-golgi intermediate compartment protein 53 FMFD1 Gp58 intracellular mannose specific lectin Intracellular mannose-specific lectin MR60 Lectin mannose-binding 1 lectin mannose-binding 1 MCFD1coagulation factor V-factor VIII combined deficiency MR60 protein ERGIC-53; Gp58; Intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose-binding, 1; Protein ERGIC-53
Gene Aliases: ERGIC-53; ERGIC53; F5F8D; FMFD1; gp58; LMAN1; MCFD1; MR60
UniProt ID: (Human) P49257
Entrez Gene ID: (Human) 3998
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.