Invitrogen
Manufacturer Code:MA536210
Catalog # PIMA536210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE55-19 |
Immunogen | Synthetic peptide within N-terminal Human LMAN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610020P13Rik AI326273 AU043785 C730041J05 endoplasmic reticulum-golgi intermediate compartment protein 53 ERGIC53 ERGIC-53 ER-Golgi intermediate compartment 53 kDa protein F5F8D FMFD1 gp58 Intracellular mannose-specific lectin MR60 lectin mannose-binding 1 lectin mannose binding 1 lectin mannose-binding 1 Lman1 MCFD1 MR60 P58 Protein ERGIC-53; endoplasmic reticulum-golgi intermediate compartment protein 53; ER-Golgi intermediate compartment 53 kDa protein; Gp58; Intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose-binding, 1; p58; Protein ERGIC-53
Gene Aliases: 2610020P13Rik; AI326273; AU043785; C730041J05; ERGIC-53; ERGIC53; F5F8D; FMFD1; gp58; LMAN1; MCFD1; MR60; P58
UniProt ID: (Human) P49257, (Mouse) Q9D0F3, (Rat) Q62902
Entrez Gene ID: (Human) 3998, (Mouse) 70361, (Rat) 116666
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.