Novus Biologicals
Manufacturer Code:NBP219366
Catalog # NBP219366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human LMAN1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum-golgi intermediate compartment protein 53; ER-Golgi intermediate compartment 53 kDa protein; ERGIC-53 ERGIC53ER-Golgi intermediate compartment 53 kDa protein F5F8Dendoplasmic reticulum-golgi intermediate compartment protein 53 FMFD1 Gp58 intracellular mannose specific lectin Intracellular mannose-specific lectin MR60 Lectin mannose-binding 1 lectin mannose-binding 1 MCFD1coagulation factor V-factor VIII combined deficiency MR60 protein ERGIC-53; Gp58; Intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose-binding, 1; Protein ERGIC-53
Gene Aliases: ERGIC-53; ERGIC53; F5F8D; FMFD1; gp58; LMAN1; MCFD1; MR60
UniProt ID: (Human) P49257
Entrez Gene ID: (Human) 3998
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.