Invitrogen
Manufacturer Code:PA5109920
Catalog # PIPA5109920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 620-819 of mouse LLGL2 (NP_663413.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130006H11Rik HGL Hugl-2 Lethal giant larvae-like protein 2 Lethal(2) giant larvae protein homolog 2 LGL2 LLGL scribble cell polarity complex component 2 Llgl2 Llglh2 RGD1560307; HGL; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 2; LLGL scribble cell polarity complex component 2
Gene Aliases: 9130006H11Rik; HGL; Hugl-2; LGL2; LLGL2; Llglh2; RGD1560307
UniProt ID: (Human) Q6P1M3, (Mouse) Q3TJ91
Entrez Gene ID: (Human) 3993, (Rat) 360661, (Mouse) 217325
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.