Novus Biologicals
Manufacturer Code:NBP181753
Catalog # NBP181753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TKYGQGFYLISPSEFERFSLSTKWLVEPRCLVDSAETKNHRPGNGAGPKKAPSRARNSGTQSDGEEKQPGLVMERALLSDERVLKEIQSTLEGDRGSGNWRSHRAAVGCSLSNGGAE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HGL; HGLlethal giant larvae (Drosophila) homolog 2 human giant larvae homolog lethal giant larvae homolog 2 (Drosophila) lethal(2) giant larvae protein homolog 2 LGL2; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal(2) giant larvae protein homolog 2; LLGL scribble cell polarity complex component 2
Gene Aliases: HGL; Hugl-2; LGL2; LLGL2
UniProt ID: (Human) Q6P1M3
Entrez Gene ID: (Human) 3993
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.